3-Bromo-5-methoxy-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

96991-1G 96991-25G 96991-500MG 96991-5G
76918-222EA 149.59 USD
76918-222 76918-224 76918-226 76918-228
3-Bromo-5-methoxy-2-methylphenol ≥97%
3-Bromo-5-methoxy-2-methylphenol
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
CAS Number: 65841-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR