3-Chloro-2-isopropoxy-6-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

88743-1G 88743-500MG 88743-5G 88743-25G
76918-170EA 1470.41 USD
76918-170 76918-172 76918-174 77660-492
3-Chloro-2-isopropoxy-6-methylbenzoic acid ≥97%
3-Chloro-2-isopropoxy-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34817853
CAS Number: 2921743-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR