3-Chloro-2-ethoxy-6-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

88722-1G 88722-500MG 88722-5G
76918-160EA 1033.32 USD
76918-160 76918-162 76918-164
3-Chloro-2-ethoxy-6-methylbenzoic acid ≥97%
3-Chloro-2-ethoxy-6-methylbenzoic acid
Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
CAS Number: 2387274-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR