3,5-Dichloro-4-methoxy-2-methylbenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

88450-1G 88450-500MG 88450-5G 88450-25G
76918-148EA 996.19 USD
76918-148 76918-150 76918-152 77657-008
3,5-Dichloro-4-methoxy-2-methylbenzoic acid ≥97% research grade
3,5-Dichloro-4-methoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34771520
CAS Number: 2918874-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR