3-Bromo-5-(9H-carbazol-9-yl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

88293-1G 88293-25G 88293-500MG 88293-5G
76918-134EA 155.96 USD
76918-134 76918-136 76918-138 76918-140
3-Bromo-5-(9H-carbazol-9-yl)benzoic acid ≥97%
3-Bromo-5-(9H-carbazol-9-yl)benzoic acid
Formula: C₁₉H₁₂BrNO₂
Storage Temperature: Ambient
CAS Number: 2921742-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR