3-(9H-carbazol-9-yl)-5-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

88289-1G 88289-500MG 88289-5G 88289-25G
76918-124EA 979.22 USD
76918-124 76918-126 76918-128 77657-006
3-(9H-carbazol-9-yl)-5-chlorobenzoic acid ≥97%
3-(9H-carbazol-9-yl)-5-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34817775
CAS Number: 2921797-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR