3-Fluoro-2-iodo-4-(methylthio)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

87244-1G 87244-25G 87244-500MG 87244-5G
76918-096EA 285.39 USD
76918-096 76918-098 76918-100 76918-102
3-Fluoro-2-iodo-4-(methylthio)benzaldehyde ≥97% research grade
3-Fluoro-2-iodo-4-(methylthio)benzaldehyde
Formula: C₈H₆FIOS
MW: 296.101 g/mol
Storage Temperature: Ambient
CAS Number: 2918918-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR