3-Chloro-5-ethoxy-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

85399-25G 85399-5G
76918-084EA 516.66 USD
76918-084 76918-086
3-Chloro-5-ethoxy-4-hydroxybenzaldehyde ≥97%
3-Chloro-5-ethoxy-4-hydroxybenzaldehyde
Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01169245
CAS Number: 70842-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR