3-Bromo-2-fluoro-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

82262-1G 82262-25G 82262-500MG 82262-5G
76918-050EA 224.92 USD
76918-050 76918-052 76918-054 76918-056
3-Bromo-2-fluoro-6-methylphenol ≥97%
3-Bromo-2-fluoro-6-methylphenol
Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Ambient
CAS Number: 1807088-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR