3-Chloro-2-fluoro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77879-1G 77879-25G 77879-500MG 77879-5G
76918-020EA 296 USD
76918-020 76918-022 76918-024 76918-026
3-Chloro-2-fluoro-6-methoxybenzoic acid ≥97%
3-Chloro-2-fluoro-6-methoxybenzoic acid
Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
CAS Number: 1782016-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR