3-Iodo-5-methylbenzonitrile ≥97%
Supplier: AOBChem USA
76647-1G
76647-25G
76647-500MG
76647-5G
76918-006EA
245.07
USD
76918-006
76918-008
76918-010
76918-012
3-Iodo-5-methylbenzonitrile ≥97%
3-Iodo-5-methylbenzonitrile
Formula:
C₈H₆IN |
CAS Number:
52107-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...