4,6-Dibromo-3-chloro-2-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

75944-1G 75944-25G 75944-500MG 75944-5G
76917-976EA 126.25 USD
76917-976 76917-978 76917-980 76917-982
4,6-Dibromo-3-chloro-2-fluoroaniline ≥97%
4,6-Dibromo-3-chloro-2-fluoroaniline
Formula: C₆H₃Br₂ClFN
MW: 303.3541 g/mol
Storage Temperature: Ambient
CAS Number: 1820717-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR