4-(2-Bromopyridin-3-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74685-1G 74685-25G 74685-500MG 74685-5G
76917-960EA 119.89 USD
76917-960 76917-962 76917-964 76917-966
4-(2-Bromopyridin-3-yl)benzaldehyde ≥97%
4-(2-Bromopyridin-3-yl)benzaldehyde
Formula: C₁₂H₈BrNO
Storage Temperature: Ambient
CAS Number: 2921896-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR