3-Bromo[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-[1,1'-biphenyl]-4-ol

72249-25G 72249-5G
76917-904EA 418 USD
76917-904 76917-906
3-Bromo[1,1'-biphenyl]-4-ol ≥97%
3-Bromo[1,1'-biphenyl]-4-ol
Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053297
CAS Number: 92-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR