3-Bromo-2,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

71119-25G 71119-5G
76917-894EA 573.95 USD
76917-894 76917-896
3-Bromo-2,5-difluorobenzaldehyde ≥97%
3-Bromo-2,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26401549
CAS Number: 112279-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR