3-(Difluoromethoxy)-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69163-1G 69163-25G 69163-500MG 69163-5G
76917-866EA 381.93 USD
76917-866 76917-868 76917-870 76917-872
3-(Difluoromethoxy)-5-methylbenzoic acid ≥97%
3-(Difluoromethoxy)-5-methylbenzoic acid
Formula: C₉H₈F₂O₃
Storage Temperature: Ambient
CAS Number: 1427403-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR