3-Fluoro-4-hydroxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

68490-1G 68490-500MG 68490-5G
76917-842EA 678.98 USD
76917-842 76917-844 76917-846
3-Fluoro-4-hydroxy-2-methylbenzoic acid ≥97%
3-Fluoro-4-hydroxy-2-methylbenzoic acid
Formula: C₈H₇FO₃
Storage Temperature: Refrigerator
CAS Number: 2386559-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR