3-Methoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

67915-1G 67915-25G 67915-500MG 67915-5G
76917-826EA 338.43 USD
76917-826 76917-828 76917-830 76917-832
3-Methoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3-Methoxy-[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₄H₁₂O₃
MW: 228.243 g/mol
CAS Number: 175153-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR