3'-Bromo-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

67729-1G 67729-25G 67729-5G
76917-816EA 79.57 USD
76917-816 76917-818 76917-820
3'-Bromo-[1,1'-biphenyl]-4-carboxylic acid ≥97% research grade
3'-Bromo-[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₃H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD08701660
CAS Number: 5737-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR