3-Bromo-5-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

67469-1G 67469-25G 67469-500MG 67469-5G
76917-802EA 228.1 USD
76917-802 76917-804 76917-806 76917-808
3-Bromo-5-fluoro-1,1'-biphenyl ≥97%
3-Bromo-5-fluoro-1,1'-biphenyl
Formula: C₁₂H₈BrF
MW: 251.094 g/mol
Storage Temperature: Ambient
CAS Number: 136649-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR