4-Bromo-2-chlorobenzal fluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-chloro-1-(difluoromethyl)benzene

66273-1G 66273-25G 66273-500MG 66273-5G
76917-788EA 422.24 USD
76917-788 76917-790 76917-792 76917-794
4-Bromo-2-chlorobenzal fluoride ≥97%
4-Bromo-2-chlorobenzal fluoride
Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391084
CAS Number: 1254034-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR