3-Bromo-4-chloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65902-1G 65902-500MG 65902-5G
76917-776EA 524.09 USD
76917-776 76917-778 76917-780
3-Bromo-4-chloro-2-fluorobenzaldehyde ≥97%
3-Bromo-4-chloro-2-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28733866
CAS Number: 1806838-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR