3-Chloro-5-formylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

65106-1G 65106-25G 65106-500MG 65106-5G
76917-732EA 206.88 USD
76917-732 76917-734 76917-736 76917-738
3-Chloro-5-formylbenzoic acid ≥95%
3-Chloro-5-formylbenzoic acid
Formula: C₈H₅ClO₃
MW: 184.5765 g/mol
Storage Temperature: Ambient
CAS Number: 153203-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR