3-Ethoxy-4,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64005-1G 64005-25G 64005-500MG 64005-5G
76917-706EA 334.19 USD
76917-706 76917-708 76917-710 76917-712
3-Ethoxy-4,5-difluorobenzaldehyde ≥97%
3-Ethoxy-4,5-difluorobenzaldehyde
Formula: C₉H₈F₂O₂
MW: 186.1554 g/mol
Storage Temperature: Ambient
CAS Number: 883535-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR