3-Bromo-2,4-difluoro-1,5-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

63710-25G 63710-5G
76917-694EA 584.56 USD
76917-694 76917-696
3-Bromo-2,4-difluoro-1,5-dimethoxybenzene ≥95%
3-Bromo-2,4-difluoro-1,5-dimethoxybenzene
Formula: C₈H₇BrF₂O₂
MW: 253.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24539372
CAS Number: 1700265-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR