3-Bromo-5-chloro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62993-1G 62993-25G 62993-500MG 62993-5G
76917-668EA 431.79 USD
76917-668 76917-670 76917-672 76917-674
3-Bromo-5-chloro-2-methylbenzoic acid ≥97%
3-Bromo-5-chloro-2-methylbenzoic acid
Formula: C₈H₆BrClO₂
Storage Temperature: Ambient
CAS Number: 1403597-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR