3-Ethoxy-2,4-difluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
62418-1G
62418-25G
62418-5G
76917-640EA
66.84
USD
76917-640
76917-642
76917-644
3-Ethoxy-2,4-difluorobenzoic acid ≥97%
3-Ethoxy-2,4-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09258723 CAS Number: 1017779-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...