3-Bromo-4-methyl-5-nitrobenzoic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
62150-25G
62150-5G
76917-630EA
463.62
USD
76917-630
76917-632
3-Bromo-4-methyl-5-nitrobenzoic acid ≥97% research grade
3-Bromo-4-methyl-5-nitrobenzoic acid
Formula:
C₈H₆BrNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD02041260 CAS Number: 34545-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...