3,5-Dibromo-7-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61887-25G 61887-5G
76917-618EA 595.17 USD
76917-618 76917-620
3,5-Dibromo-7-methyl-1H-indazole ≥97%
3,5-Dibromo-7-methyl-1H-indazole
Formula: C₈H₆Br₂N₂
Storage Temperature: Ambient
CAS Number: 1510788-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR