3-Bromo-6-chloro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

61880-1G 61880-25G 61880-500MG 61880-5G
76917-606EA 126.25 USD
76917-606 76917-608 76917-610 76917-612
3-Bromo-6-chloro-1H-indazole ≥95%
3-Bromo-6-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781578
CAS Number: 885521-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR