3,6-Dibromo-4-fluoro-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
61877-1G
61877-500MG
76917-594EA
1840.67
USD
76917-594
76917-596
3,6-Dibromo-4-fluoro-1H-indazole ≥97%
3,6-Dibromo-4-fluoro-1H-indazole
Formula:
C₇H₃Br₂FN₂ Storage Temperature: Refrigerator |
CAS Number:
887567-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...