3,4-Dibromo-1H-indazole ≥95% research grade

Supplier: AOBChem USA

Warning

61873-1G 61873-25G 61873-500MG 61873-5G
76917-580EA 301.3 USD
76917-580 76917-582 76917-584 76917-586
3,4-Dibromo-1H-indazole ≥95% research grade
3,4-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
Storage Temperature: Refrigerator
CAS Number: 885521-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR