3,7-Dibromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61871-1G 61871-25G 61871-500MG 61871-5G
76917-564EA 351.16 USD
76917-564 76917-566 76917-568 76917-570
3,7-Dibromo-1H-indazole ≥97%
3,7-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
MW: 275.928 g/mol
Storage Temperature: Ambient
CAS Number: 1541407-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR