3,5-Difluoro-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

61865-25G 61865-5G
76917-552EA 481.65 USD
76917-552 76917-554
3,5-Difluoro-N,N-dimethylaniline ≥97%
3,5-Difluoro-N,N-dimethylaniline
Formula: C₈H₉F₂N
Storage Temperature: Ambient
CAS Number: 123855-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR