3-Bromo-6-fluoro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61771-1G 61771-25G 61771-5G
76917-538EA 117.76 USD
76917-538 76917-540 76917-542
3-Bromo-6-fluoro-1H-indazole ≥97%
3-Bromo-6-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781603
CAS Number: 885522-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR