3-Chloro-2,4,6-trifluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61112-1G 61112-25G 61112-500MG 61112-5G
76917-506EA 198.39 USD
76917-506 76917-508 76917-510 76917-512
3-Chloro-2,4,6-trifluorobenzaldehyde ≥97%
3-Chloro-2,4,6-trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₂ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD11845970
CAS Number: 1160573-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR