3-Bromo-5-tert-butyl-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-(tert-butyl)-2-hydroxybenzaldehyde

60883-1G 60883-25G 60883-500MG 60883-5G
76917-490EA 241.89 USD
76917-490 76917-492 76917-494 76917-496
3-Bromo-5-tert-butyl-2-hydroxybenzaldehyde ≥97%
3-Bromo-5-tert-butyl-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739522
CAS Number: 119646-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR