3-Iodo-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

60623-1G 60623-500MG 60623-5G
76917-476EA 530.45 USD
76917-476 76917-478 76917-480
3-Iodo-5-(trifluoromethyl)benzaldehyde ≥97%
3-Iodo-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃IO
MW: 300.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977391
CAS Number: 868166-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR