3-Hydroxy-5-(trifluoromethoxy)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

58229-1G 58229-25G 58229-500MG 58229-5G
76917-424EA 155.96 USD
76917-424 76917-426 76917-428 76917-430
3-Hydroxy-5-(trifluoromethoxy)benzaldehyde ≥97% research grade
3-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Formula: C₈H₅F₃O₃
MW: 206.1187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397429
CAS Number: 1261852-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR