3-Isopropoxy-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-2-isopropoxyanisole, 4-Methoxy-3-[(prop-2-yl)oxy]benzoic acid

58206-1G 58206-25G 58206-5G
76917-412EA 88.06 USD
76917-412 76917-414 76917-416
3-Isopropoxy-4-methoxybenzoic acid ≥97%
3-Isopropoxy-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00831064
CAS Number: 159783-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR