4-(Benzyloxy)-3-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57997-1G 57997-500MG 57997-5G
76917-400EA 527.27 USD
76917-400 76917-402 76917-404
4-(Benzyloxy)-3-fluoro-5-formylbenzoic acid ≥97%
4-(Benzyloxy)-3-fluoro-5-formylbenzoic acid
Formula: C₁₅H₁₁FO₄
Storage Temperature: Refrigerator
CAS Number: 2757730-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR