3-Bromo-N-methoxy-N-methyl-5-nitrobenzamide ≥95%

Supplier: AOBChem USA

Warning

57866-1G 57866-25G 57866-5G
76917-388EA 79.57 USD
76917-388 76917-390 76917-392
3-Bromo-N-methoxy-N-methyl-5-nitrobenzamide ≥95%
3-Bromo-N-methoxy-N-methyl-5-nitrobenzamide
Formula: C₉H₉BrN₂O₄
MW: 289.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25132888
CAS Number: 1519917-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR