4,5-Dichloro-o-toluidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Dichloro-2-methylaniline

57860-25G 57860-5G
76917-376EA 346.92 USD
76917-376 76917-378
4,5-Dichloro-o-toluidine ≥97%
4,5-Dichloro-o-toluidine
Formula: C₇H₇Cl₂N
MW: 176.0432 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855825
CAS Number: 2387-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR