3-Methyl-5-(methylthio)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

57473-1G 57473-25G 57473-500MG 57473-5G
76917-342EA 154.9 USD
76917-342 76917-344 76917-346 76917-348
3-Methyl-5-(methylthio)benzoic acid ≥97% research grade
3-Methyl-5-(methylthio)benzoic acid
Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD22489512
CAS Number: 861560-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR