4-(Benzyloxy)-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

57375-1G 57375-25G 57375-500MG 57375-5G
76917-330EA 242.95 USD
76917-330 76917-332 76917-334 76917-336
4-(Benzyloxy)-2-fluoropyridine ≥97%
4-(Benzyloxy)-2-fluoropyridine
Formula: C₁₂H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD16658936
CAS Number: 1253792-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR