4-Bromo-1,5-dimethyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Bromo-1,5-dimethylpyrazole
57065-25G
57065-5G
76917-310EA
495.45
USD
76917-310
76917-312
4-Bromo-1,5-dimethyl-1H-pyrazole ≥97%
4-Bromo-1,5-dimethyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₇BrN₂ MW: 175.0265 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09759006 CAS Number: 5775-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...