3-Bromo-6-fluoro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57018-1G 57018-25G 57018-500MG 57018-5G
76917-296EA 393.6 USD
76917-296 76917-298 76917-300 76917-302
3-Bromo-6-fluoro-2-hydroxybenzoic acid ≥97%
3-Bromo-6-fluoro-2-hydroxybenzoic acid
Formula: C₇H₄BrFO₃
Storage Temperature: Ambient
CAS Number: 1538235-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR