4-(Benzyloxy)-2-bromo-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56806-1G 56806-25G 56806-500MG 56806-5G
76917-276EA 213.25 USD
76917-276 76917-278 76917-280 76917-282
4-(Benzyloxy)-2-bromo-1-iodobenzene ≥97%
4-(Benzyloxy)-2-bromo-1-iodobenzene
Formula: C₁₃H₁₀BrIO
MW: 389.026 g/mol
Storage Temperature: Ambient
CAS Number: 2140305-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR