4-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56171-1G 56171-25G 56171-500MG 56171-5G
76917-246EA 340.55 USD
76917-246 76917-248 76917-250 76917-252
4-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde ≥97%
4-(1-Methyl-1H-pyrazol-5-yl)benzaldehyde
Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD8669907
CAS Number: 179055-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR