3-Propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56037-25G 56037-5G
76917-236EA 279.02 USD
76917-236 76917-238
3-Propoxybenzaldehyde ≥97%
3-Propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01993679
CAS Number: 67698-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR